Ethyl-3-methyl-3-phenylglycidate (mixture of cis and trans isomers) ≥98.0%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Ethyl 3-Methyl-3-phenylglycidate (mixture of isomers), Ethylmethylphenylglycidate Ethyl 3-methyl-3-phenylglycidate 3-Methyl-3-phenyl glycidic acid ethyl ester, Ethyl 2,3-Epoxy-3-phenylbutyrate (mixture of isomers), 2,3-Epoxy-3-phenylbutyric acid Ethyl Ester (mixture of isomers), 3-Methyl-3-phenylglycidic acid Ethyl Ester (mixture of isomers)

E1057-500GM E1057-100GM
700006-620EA 709.55 USD
700006-620 700006-618
Ethyl-3-methyl-3-phenylglycidate (mixture of cis and trans isomers) ≥98.0%
Ethyl 3-methyl-3-phenylglycidate (mixture of cis and trans isomers)

Ethyl Methylphenylglycidate is used as a perfumery ingredient with fresh, fruity notes.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₂H₁₄O₃
MW: 206.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00022338
CAS Number: 77-83-8

Order Now

Specification Test Results

Assay 98.0 %
Acid Value max. 2.0
Specific Gravity @ 20D/20°C 1.086 - 1.096
Refractive Index @ 20°C 1.504 - 1.513

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR