Ethyl cellulose FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Ethylcellulose
Ethyl Cellulose, FCC, a derivative of cellulose, is often used as an emulsifier food additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Storage Temperature:
Ambient |
MDL Number:
MFCD00131037 CAS Number: 9004-57-3 |
Specification Test Results
Assay (Ethoxy Groups; Dried Basis) | 44.0 - 50.0% |
Viscosity | To Pass Test |
Lead (Pb) | max. 3 mg/kg |
Loss On Drying | max. 3% |
Residue On Ignition | max. 0.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...