Ethyl-2-cyanoacrylate
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
Ethyl 2-cyanoacrylate
E2875-1GM
E2875-5GM
E2875-10GM
700007-974EA
187.25
USD
700007-974
700007-976
700007-972
Ethyl-2-cyanoacrylate
Ethyl-2-cyanoacrylate
Ethyl 2-cyanoacrylate is an ethyl ester of 2-cyano-2-propenoic acid and can be used for gluing of several types of materials.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₆H₇NO₂ MW: 125.1253 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00045615 CAS Number: 7085-85-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...