Elemi gum
Supplier: Spectrum Chemical Mfg. Corp.
Elemi Gum is used as a plasticizer and film-former.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Storage Temperature:
Ambient |
MDL Number:
MFCD01771924 CAS Number: 9000-75-3 |
Specification Test Results
Melting Point | 80 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...