D-ɑ-Tocopherol succinate ≥96.0% USP
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Vitamin E Succinate, D-α-Tocopherol Succinate, 4-Oxo-4-(((R)-2,5,7,8-tetramethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-yl)oxy)butanoic acid
V1035-1KG
V1035-25GM
V1035-500GM
V1035-100GM
95036-562EA
1938.65
USD
95036-562
95036-564
95036-566
95036-560
D-ɑ-Tocopherol succinate ≥96.0% USP
D-alpha-Tocopherol succinate
RRR-Vitamin E Succinate, 1 mg = 0.81 alpha-tocopherol equivalents, USP.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₃₃H₅₄O₅ MW: 530.79 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00072055 CAS Number: 4345-03-3 |
Specification Test Results
Assay | 96.0 - 102.0 % |
Acidity | To Pass Test |
Identification | To Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...