Sodium-D-pantothenate
Supplier: Spectrum Chemical Mfg. Corp.
D-Pantothenic Acid Sodium Salt is also known as pantothenate and is soluble in water. It is considered an essential nutrient for several types of animals. It is also an ingredient in some skin or hair products.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₉H₁₇NO₅ Storage Temperature: Ambient |
MDL Number:
MFCD00002767 CAS Number: 867-81-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...