D-Pantethine ≥98%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (2R)-N-[3-[2-[2-[3-[[(2R)-2,4-Dihydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyldisulphanyl]ethylamino]-3-oxopropyl]-2,4-dihydroxy-3,3-dimethylbutanamid

P1024-5GM P1024-1GM P1024-100GM P1024-25GM
700008-172EA 249.02 USD
700008-172 700008-168 700008-166 700008-170
D-Pantethine ≥98%
D-Pantethine

D-Pantethine, also known as Bis-pantethine, can be used to treat acne and improve the blood-cholesterol profile.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₂H₄₂N₄O₈S₂
MW: 554.72 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00151211
CAS Number: 16816-67-4

Order Now

Specification Test Results

Assay (Dried Basis) 98 % min.
Water 18 - 22 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR