DL-Methionine ≥98.5% FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
2-Amino-4-(methylthio)butanoic acid, (±)-2-Amino-4-(methylthio)butyric acid, Methioninum racemicum, Methionine racematic, H-DL-Met-OH, (±)-Methionine, (±)-2-Amino-4-(methylthio)butanoic acid
DL-Methionine, FCC is used as an amino acid nutritional supplement and flavor enhancer. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₅H₁₁NO₂S MW: 149.21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00063096 CAS Number: 59-51-8 |
Specification Test Results
Assay (C₅H₁₁No₂S) | 98.5 - 101.5 % |
Lead (Pb) | max. 5 mg/kg |
Loss On Drying | max. 0.5 % |
Residue On Ignition | max. 0.1 % |
Identification | To Pass Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...