Lactic acid ≥88%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: DL-Lactic acid, 2-Hydroxypropionic acid, 2-Hydroxypropanoic acid

L2016-2.5LT L2016-500ML
10841-786EA 405.65 USD
10841-786 10841-788
Lactic acid ≥88%
Lactic acid

DL-Lactic Acid, Racemic, also known as milk acid, is used in various biochemical processes. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.


GHS hazard : Corrosion
UN3265

Formula: H₃CCH(OH)COOH
MW: 90.08 g/mol
Density: 1.209
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS Number: 50-21-5
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Assay Approx. 88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR