Dipropylene glycol methyl ether acetate ≥98%
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
Di(propyleneglycol)methyletheracetate
D1111-20LT
D1111-1LT
D1111-250ML
D1111-4LTGL
700004-454EA
1380
USD
700004-454
700004-452
700004-456
700004-458
Dipropylene glycol methyl ether acetate ≥98%
Dipropylene glycol methyl ether acetate
Dipropylene Glycol Methyl Ether Acetate is made up of three isomeric chemical compounds. It is a colorless liquid with out any discernible odor. It can be used as a plasticizer or as a solvent. It is considered an ideal additive for skin, hair, and perfume products.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₉H₁₈O₅ MW: 190.2300 g/mol Density: 0.970 Storage Temperature: Ambient |
MDL Number:
MFCD00144160 CAS Number: 88917-22-0 |
Specification Test Results
Assay | 98 % |
Color (APHA) | max.15 |
Acidity (As Ch₃COOH) | max. 0.05 % |
Water | max. 0.05 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...