1,3-Dihydroxyacetone dimer ≥98%
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
2,5-Dihydroxy-1,4-dioxane-2,5-dimethanol
D1038-100GM
D1038-1KG
D1038-25GM
D1038-25KGBL
700004-046EA
98.35
USD
700004-046
700004-048
700004-050
700004-052
1,3-Dihydroxyacetone dimer ≥98%
1,3-Dihydroxyacetone dimer
Dihydroxyacetone, Dimer, also known as glycerone, is used as an ingredient in sunless tanning products and can be derived from the fermentation of glycerin.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₆H₁₂O₆ MW: 180.16 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00051019 CAS Number: 62147-49-3 |
Specification Test Results
Assay | 98 % min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...