Pentasodium diethylenetriaminepentaacetate solution 41% in water

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Diethylenetriaminepentaacetic Acid Pentasodium Salt

D1156-100ML D1156-500ML D1156-4LT
700014-012EA 89.31 USD
700014-012 700014-016 700014-014
Pentasodium diethylenetriaminepentaacetate solution 41% in water
Pentasodium diethylenetriaminepentaacetate

Diethylenetriaminepentaacetic Acid, Pentasodium Salt, 41 Percent Solution in Water is also known under the abbreviation DETA and is a colorless liquid that is hygroscopic and soluble in both water and organic solvent that are polarized. Used as a tridentate ligand forming complex in coordination chemistry, it is also used for the extraction of gases that are acid in the oil industry

Formula: C₁₄H₂₃N₃O₁₀
MW: 503.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00051016
CAS Number: 140-01-2

Order Now

Specification Test Results

Assay 39 - 41 %
pH of a 1% Solution 11.0 - 12.0
Color (APHA) max. 250

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR