D-(+)-Glucurono-6,3-lactone
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
(R)-2-((2S,3R,4S)-3,4-Dihydroxy-5-oxotetrahydrofuran-2-yl)-2-hydroxyacetaldehyde, D-(+)-Glucuronolactone
G2160-100GM
G2160-500GM
G2160-25GM
700008-376EA
128.9
USD
700008-376
700008-382
700008-378
D-(+)-Glucurono-6,3-lactone
D(+)-Glucurono-6,3-lactone
D-Glucurono-3,6-lactone, also known as glucuronolactone, is a natural chemical that is a component of almost all connective tissues. It is used as a precursor for ascorbic acid synthesis.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₆H₈O₆ MW: 176.12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00135622 CAS Number: 32449-92-6 |
Specification Test Results
Appearance | White Powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...