D-Glucoronic acid

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: D-(+)Glucuronic acid, D-Glucuronic acid, D-(+)-glucuronic acid, (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid

G2028-100GM G2028-25GM G2028-2KGBL G2028-5GM
700009-510EA 848 USD
700009-510 700009-512 700009-514 700009-516
D-Glucoronic acid
D-Glucoronic acid

D-Glucuronic Acid, also known as β-D-glucopyranuronic acid, is a carboxylic acid with a structure similar to glucose and used in the determination of urinary steroids and of steroid conjugates in blood.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₆H₁₀O₇
Storage Temperature: Refrigerator
MDL Number: MFCD00166981
CAS Number: 6556-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR