D-(+)-Glucosamine hydrochloride ≥98%
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
2-Amino-2-deoxy-D-glucopyranose Hydrochloride, D-Glucosamine hydrochloride, (2R,3R,4S,5R)-2-Amino-3,4,5,6-tetrahydroxyhexanal hydrochloride
D-Glucosamine Hydrochloride is an amino sugar and plays an important role in the biochemical synthesis of glycosylated lipids and proteins. It is most commonly used as a non-vitamin dietary supplement.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₆H₁₃NO₅ MW: 215.63 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00135831 CAS Number: 66-84-2 |
Specification Test Results
Assay | 98 % min. |
Moisture | max.1.5 % |
Heavy Metals | max. 10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...