Acid red 33

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: D&C Red No. 33, Disodium (3Z)-5-amino-4-oxo-3-(phenylhydrazinylidene)naphthalene-2,7-disulphonate, Azo Fuchsine

DC145-25GM DC145-100GM DC145-500GM DC145-5KGBL
700014-176EA 242.71 USD
700014-176 700014-174 700014-178 700014-180
Acid red 33
Acid red 33

D and C Red No. 33 | Non-graded D and C Red No. 33 supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


GHS hazard : Exclamation mark

Formula: C₁₆H₁₁N₃Na₂O₇S₂
MW: 467.378 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00071877
CAS Number: 3567-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR