Orange II
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
Tropaeolin OOO-2, acid orange 7, acid orange A, D and C Orange No. 4, 4-(2-Hydroxy-1-naphthylazo)benzenesulphonic acid sodium salt, Orange II sodium salt, p-(2-Hydroxy-1-naphthylazo)benzenesulphonic acid sodium salt, Gold orange
D1130-500GM
D1130-100GM
700010-014EA
1546.7
USD
700010-014
700010-010
Orange II
Orange II
D and C Orange No. 4 | Non-graded D and C Orange No. 4 supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
GHS hazard : Exclamation mark
Formula:
C₁₆H₁₁N₂NaO₄S MW: 350.32 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011657 CAS Number: 633-96-5 |
Specification Test Results
Identification | To Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...