Alizarin cyanin green F

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: acid Green 25, D and C Green No. 5, Acid Green 25, Alizarin Cyanin Green F

D1119-25GM D1119-500GM D1119-100GM
700010-006EA 93.6 USD
700010-006 700010-008 700010-004
Alizarin cyanin green F
Alizarin cyanin green F

D and C Green No. 5 | Non-graded D and C Green No. 5 supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₈H₂₂N₂O₈S₂
MW: 622.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001193
CAS Number: 4403-90-1

Order Now

Specification Test Results

Identification To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR