Cyclopentanone ≥99%
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
C1124-100ML
C1124-4LT
C1124-500ML
10841-766EA
88.2
USD
10841-766
10841-768
10841-770
Cyclopentanone ≥99%
Cyclopentanone
Cyclopentanone is a liquid that has an odor similar to peppermint and is devoid of color. It's an organic compound and a cyclic ketone that most often used as a fragrance.
GHS hazard : Flame;Exclamation mark
UN2245
Formula:
C₅H₈O MW: 84.12 g/mol Density: 0.951 Storage Temperature: Ambient |
MDL Number:
MFCD00001409 CAS Number: 120-92-3 UN: 2245 ADR: 3,III |
Specification Test Results
Assay | 99 % min. |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...