Cyclohexyl methacrylate ≥97.0%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Methacrylic Acid Cyclohexyl Ester

C1687-100GM C1687-500GM
700005-374EA 124.8 USD
700005-374 700005-376
Cyclohexyl methacrylate ≥97.0%
Cyclohexyl methacrylate

Cyclohexyl Methacrylate is a cycloalkane monomer used in the industrial use manufacture of thermoplastics .


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₀H₁₆O₂
MW: 168.24 g/mol
Density: 0.964
Storage Temperature: Ambient
MDL Number: MFCD00014292
CAS Number: 101-43-9

Order Now

Specification Test Results

Assay (GC) 97.0% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR