Cyanoacetic acid ≥98%

Supplier: SPECTRUM CHEMICAL MFG CORP

C2322-100GM C2322-500GM
10841-756EA 78 USD
10841-756 10841-758
Cyanoacetic acid ≥98%
Cyanoacetic acid

Cyanoacetic Acid, also known as 2-cyanoacetic acid, is an organic compound and precursor to cyanoacrylates and components of adhesives. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.


GHS hazard : Corrosion;Exclamation mark
UN1759

Formula: C₃H₃NO₂
MW: 85.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002677
CAS Number: 372-09-8
UN: 1759
ADR: 8,III

Order Now

Specification Test Results

Assay 98 % min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR