Coconut oil, refined
Supplier: SPECTRUM CHEMICAL MFG CORP
CO110-2.5KG
CO110-200LTB
CO110-500GM
CO110-12KG
700011-706EA
263.8
USD
700011-706
700011-708
700011-710
700011-704
Coconut oil, refined
Coconut oil
Coconut Oil, Refined is taken from the meat of coconuts that come from the coconut palm. It is considered an edible oil and has several purposes in food, industry, and medicine. It contains high saturated fat.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Storage Temperature:
Ambient |
MDL Number:
MFCD00130817 CAS Number: 8001-31-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...