Acid Red 18 (Food Red No. 102)

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Cochineal Red A, New Coccine, Scarlet 3R, Sodium 7-hydroxy-8-((4-sulphonatonaphthalen-1-yl)diazenyl)naphthalene-1,3-disulphonate, Ponceau 4R, Cochenille red

CO102-25GM CO102-50KGBL CO102-100GM
700009-996EA 70.55 USD
700009-996 700009-998 700009-994
Acid Red 18 (Food Red No. 102)
Acid Red 18 (Food Red No. 102)

Cochineal Red A, also known as ponceau 4R, is typically synthesized from aromatic hydrocarbons and used as a synthetic food coloring. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₀H₁₄N₂O₁₀S₃
MW: 604.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004084
CAS Number: 2611-82-7

Order Now

Specification Test Results

Absorption Max 506 (335) NM

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR