Clove oil FCC

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Oils, clove leaf, Eugenia caryophyllata leaf oil

CL130-500MLG CL130-125ML CL130-25ML
700010-602EA 607.75 USD
700010-602 95032-652 95032-654
Clove oil FCC
Clove oil

Clove Oil, FCC is a flavor and fragarant ingredient. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Boiling Pt: 251 °C (484 °F)
Density: 1.04
Flash Pt: 115 °C (239 °F)
Storage Temperature: Ambient
MDL Number: MFCD00130815
CAS Number: 8000-34-8

Order Now

Specification Test Results

Assay (Phenols) 85.0 % min.
Angular Rotation -1.5° to 0°
Refractive Index @ 20°C 1.527 - 1.535
Solubility in 70% Alcohol To Pass Test
Specific Gravity @ 25°C 1.038 - 1.060
Phenols To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR