Cistus oil

Supplier: SPECTRUM CHEMICAL MFG CORP

C1842-100ML C1842-25ML C1842-5ML
700013-874EA 751.65 USD
700013-874 700013-876 700013-878
Cistus oil
Cistus oil

Cistus Oil is a fragrance ingredient for perfumery and cosmetics.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
CAS Number: 8016-26-0

Order Now

Specification Test Results

Appearance Light Brown to V


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR