Cinnamyl acetate ≥95%
Supplier: SPECTRUM CHEMICAL MFG CORP
C2765-1LT
C2765-4LT
700004-444EA
276.8
USD
700004-444
700004-448
Cinnamyl acetate ≥95%
Cinnamyl acetate
Cinnamyl Acetate has a very attractive, fruity, balsamic aroma.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₁H₁₂O₂ MW: 176.22 g/mol Density: 1.057 Storage Temperature: Ambient |
MDL Number:
MFCD00008722 CAS Number: 103-54-8 |
Specification Test Results
Assay | 95 % min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...