Choline dihydrogen citrate ≥98%
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
Choline dihydrogencitrate salt, 2-Hydroxy-N,N,N-trimethylethan-1-aminium 3,4-dicarboxy-3-hydroxybutanoate
C1229-2.5KG
C1229-500GM
C1229-50KGBL
C1229-12KG
700006-180EA
683.8
USD
700006-180
700006-182
700006-184
700006-178
Choline dihydrogen citrate ≥98%
Choline dihydrogen citrate
Choline Dihydrogen Citrate is part of a class of quaternary ammonium salts containing the N,N,N-trimethylethanolammonium cation and Dihydrogen Citrate counteranion. Choline is the precursor for the neurotransmitter acetylcholine, which is involved in many functions including memory and muscle control. This item is of known assay and suitable for most chemical uses.
GHS hazard : Exclamation mark
Formula:
C₁₁H₂₁NO₈ MW: 295.29 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00038734 CAS Number: 77-91-8 |
Specification Test Results
Assay | 98 % min. |
Water | max.0.25 % |
Residue On Ignition | max. 0.05 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...