Chlorophyllin, coppered trisodium salt, powder

Supplier: SPECTRUM CHEMICAL MFG CORP

C1443-100GM C1443-12KGBL C1443-500GM C1443-25GM
700013-756EA 340.7 USD
700013-756 700013-758 700013-762 700013-760
Chlorophyllin, coppered trisodium salt, powder
Chlorophyllin, coppered trisodium salt

Chlorophyllin, Powder, also known as Natural Green 3, E141, and is a water soluble salt that is a semi-synthetic derivative of chlorophyll.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₃₄H₃₁CuN₄Na₃O₆
MW: 724.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012149
CAS Number: 11006-34-1

Order Now

Specification Test Results

Content of Chelated Copper 4.0 % min.
Content of Total Copper 4.25 % min.
pH (1% in Water) 9.0 - 10.9
Ash max. 36 %
Arsenic max. 3 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR