Chlorogenic acid hydrate ≥97%
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
3-(3,4-Dihydroxycinnamoyl)quinic acid hydrate, (1S,3R,4R,5R)-3-(((3-(3,4-dihydroxyphenyl)acryloyl)oxy)-1,4,5-trihydroxycyclohexanecarboxylic acid hydrate, 3-O-Caffeoylquinic acid hydrate, (1S,3R,4R,5R)-3-[[3-(3,4-Dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-1,4,5-trihydroxycyclohexanecarboxylic acid hydrate
Chlorogenic Acid, also known as 3-(3,4-Dihydroxycinnamoyl)quinate, is a crucial intermediate in lignin biosynthesis.
Non-graded Chlorogenic Acid supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₆H₁₈O₉ MW: 354.31 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003862 CAS Number: 327-97-9 |
Specification Test Results
Assay (Anhydrous Basis) | 97 % min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...