Cetostearyl alcohol NF
Supplier: SPECTRUM CHEMICAL MFG CORP
C1888-12KG
C1888-2.5KG
C1888-45KGBL
C1888-500GM
700002-716EA
1637.35
USD
700002-716
75810-856
75810-858
75810-860
Cetostearyl alcohol NF
Cetostearyl alcohol
Cetostearyl Alcohol, NF is used as a stabilizer and opacifying agent.
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Storage Temperature:
Ambient |
MDL Number:
MFCD04113584 CAS Number: 8005-44-5 |
Specification Test Results
Stearyl Alcohol and Cetyl Alcohol | 90.0 - 102.0 % |
Stearyl Alcohol | 40.0 % min. |
Hydroxyl Value | 208 - 228 |
Acid Value | max. 2 |
Iodine Value | max. 4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...