Castor oil ethoxylated
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Polyoxyl 35 castor oil, Castor oil polyoxyl 35, PEG 35 castor oil
Castor Oil, Ethoxylated is an oil that is derived from the castor beans that has been polyethoxylated. A colorless oil that has no taste or smell and boils at 313 C, it is used in the production of many products such as lubricants, paints, dyes, waxes, perfumes, pharmaceuticals and plastics.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Density:
1.050 Storage Temperature: Ambient |
MDL Number:
MFCD00130879 CAS Number: 61791-12-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...