Canola oil

Supplier: SPECTRUM CHEMICAL MFG CORP

C1523-16KGBL C1523-2.5KG C1523-500GM
700011-668EA 705.5 USD
700011-668 700011-670 700011-674
Canola oil
Canola oil

Canola Oil is an excellent emollient and odorless vegetable oil used as a moisturizing agent for skin conditioning.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Density: 1.00
Storage Temperature: Ambient
MDL Number: MFCD00217791
CAS Number: 120962-03-0

Order Now

Specification Test Results

Appearance @ 25 °C Clear, Bright, Y
Iodine Value 94 - 126
Saponification Value 186 - 198
Acid Value max. 0.5
Peroxide Value max. 10

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR