Calcium pantothenate ≥98.0%, powder USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Calcium D-Pantothenate, D-Pantothenic acid calcium salt, D-Calcium pantothenate, Calcium (R)-3-(2,4-dihydroxy-3,3-dimethylbutanamido)propanoate, Vitamin B5 calcium salt, Calcium D-(+)-pantothenate

CA159-50KGBL CA159-2.5KG CA159-500GM CA159-12KG CA159-100GM
700011-692EA 12722.15 USD
700011-692 95032-320 95032-322 700011-690 95032-318
Calcium pantothenate ≥98.0%, powder USP
Calcium pantothenate

CALCIUM PANTOTHENATE, POWDER, USP is a water-soluble version of vitamin B5 and is often used in dietary supplements because, as a salt, it is more stable than pantothenic acid in the digestive tract and may improve oxygen utilization efficiency and reduce lactic acid accumulation in athletes. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities. Storage temperature - LIGHT SENSITIVE, HYGROSCOPIC: Keep tightly closed in light-resistant containers.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₈H₃₂CaN₂O₁₀
MW: 476.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002766
CAS Number: 137-08-6

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 102.0 %
Calcium Content (Ca; Dried Basis) 8.2 - 8.6 %
Optical Rotation +25.0° to +27.5
Loss On Drying max. 5.0 %
Alkalinity To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR