Calcium disodium EDTA hydrate 97.0-102.0% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Edetate calcium disodium hydrate

E1194-125GM E1194-500GM E1194-2.5KG
95033-044EA 159.7 USD
95033-044 95033-048 95033-046
Calcium disodium EDTA hydrate 97.0-102.0% FCC
Calcium disodium EDTA hydrate

Calcium Disodium EDTA, FCC is used as a flavor and color preservative. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Storage Temperature: Ambient
MDL Number: MFCD00149453
CAS Number: 23411-34-9

Order Now

Specification Test Results

Assay (Anhydrous Basis) 97.0 - 102.0 %
pH of a 1 in 100 Solution 6.5 - 7.5
Lead (Pb) max. 4 mg/kg
Magnesium-Chelating Substances To Pass Test
Nitrilotriacetic Acid[N[(HOCOCH₂)₃N] max. 0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR