Calcium-D-saccharate tetrahydrate
Supplier: Spectrum Chemical Mfg. Corp.
|
|
Calcium D-glucarate, Tetrahydrate is used as a clarifying agent for the removal of high molecular weight components by filtration.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₆H₈CaO₈.4H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150704 CAS Number: 5793-89-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...