Calcium ascorbate dihydrate ≥98.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Calcium (R)-5-((S)-1,2-dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one salt dihydrate, Vitamin C calcium salt, L-Ascorbic acid calcium salt dihydrate, Calcium L-ascorbate dihydrate

C1764-2.5KG C1764-100GM C1764-500GM
95032-214EA 1306.05 USD
95032-214 95032-212 95032-216
Calcium ascorbate dihydrate ≥98.0% USP
Calcium ascorbate dihydrate

Calcium Ascorbate, Dihydrate, USP is a form of vitamin C that is used to treat low levels of vitamin C. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₂H₁₄CaO₁₂·2H₂O
MW: 426.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149216
CAS Number: 5743-28-2

Order Now

Specification Test Results

Assay 98.0 - 101.0 %
Specific Rotation [A]D +95° to +97°
pH of a 1 in 10 Solution 6.8 - 7.4
Loss On Drying max. 0.1 %
Arsenic (As) max. 3 µg/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR