Butyl myristate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Myristic Acid Butyl Ester, Butyl tetradecanoate

B1264-25GM B1264-100GM
700013-608EA 177.05 USD
700013-608 700013-606
Butyl myristate
Butyl myristate

GHS hazard : Exclamation mark

Formula: C₁₈H₃₆O₂
MW: 284.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00015077
CAS Number: 110-36-1

Order Now

Specification Test Results

Appearance Colorless to Pal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR