Black pepper oil FCC

Supplier: Spectrum Chemical Mfg. Corp.

B1222-125ML B1222-500MLGL
95030-748EA 294.4 USD
95030-748 75810-686
Black pepper oil FCC
Black pepper oil

Black Pepper Oil, FCC is a food and flavor ingredient. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Flame;Exclamation mark UN1197

Storage Temperature: Ambient
MDL Number: MFCD01635834
CAS Number: 8006-82-4
UN: 1197
ADR: 3,III

Order Now

Specification Test Results

Angular Rotation -1 to -23
Refractive Index @ 20°C 1.479- 1.488
Solubility in Alcohol To Pass Test
Specific Gravity 0.864- 0.884
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR