Bifonazole

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1-(alpha,4-Diphenylbenzyl)imidazole, 1-([1,1'-Biphenyl]-4-yl(phenyl)methyl)-1H-imidazole

B3366-1GM B3366-5GM
700013-662EA 230.6 USD
700013-662 700013-666
Bifonazole
Bifonazole

Bifonazole is known as an imidazole antifungal drug.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₂H₁₈N₂
Storage Temperature: Ambient
MDL Number: MFCD00865567
CAS Number: 60628-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR