2-Hydroxy-4-methoxy-5-sulfobenzophenone ≥97.0%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Sulisobenzone, Benzophenone-4, 5-Benzoyl-4-hydroxy-2-methoxybenzenesulphonic acid, 2-Hydroxy-4-methoxybenzophenone-5-sulphonic Acid

B1082-12KGBL B1082-500GM B1082-100GM B1082-2.5KG B1082-25GM
700004-114EA 3402.6 USD
700004-114 700004-118 700004-112 700004-116 10860-440
2-Hydroxy-4-methoxy-5-sulfobenzophenone ≥97.0%
2-Hydroxy-4-methoxy-5-sulfobenzophenone

Benzophenone-4 is also known as sulisobenzone and is typically an ingredient in most sunscreens. It is used as a skin protectant from damage by short-wave UVA ultraviolet light and UVB.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₄H₁₂O₆S
MW: 308.30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00024962
CAS Number: 4065-45-6

Order Now

Specification Test Results

Assay (Dry Basis) 97.0 - 103.0 %
Extinction Coefficient in 95% Ethanol at 285 NM 460 min.
pH of a 1% Solution 1.2 - 2.2
Color (Gardner; 10% W/V in Water) max. 4.0
Solubility of 10% W/V in Water To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR