Benzenesulfonyl chloride ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Benzene sulphochloride

B2003-1LT B2003-250ML
10841-752EA 363.6 USD
10841-752 10841-754
Benzenesulfonyl chloride ≥99%
Benzenesulphonyl chloride

Benzenesulfonyl Chloride, an organosulfur compound, dissolves in organic solvents and is used to prepare sulfonate and sulfonamides esters by reactions with alcohols and amines.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₆H₅ClO₂S
MW: 176.62 g/mol
Density: 1.384
Storage Temperature: Ambient
MDL Number: MFCD00007426
CAS Number: 98-09-9
UN: 2225
ADR: 8,III

Order Now

Specification Test Results

Assay 99 % min.
Color (APHA) max. 20
Free Acid max. 0.05 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR