Sorbitan sesquioleate
Supplier: Spectrum Chemical Mfg. Corp.
Arlacel(R) C , also known as Arlacel 83, is used as an emulsifier and nonionic surfactant in pharmaceutical formulations. The primary use for this product is as an emulsifier for creams and lotions that are rich in non-polar oils and waxes.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₄₂H₇₆O₇ Density: 0.989 Storage Temperature: Ambient |
MDL Number:
MFCD00151568 CAS Number: 8007-43-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...