Diammonium hydrogen orthophosphate ≥96.0%, granular NF
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate
A1244-12KG
A1244-2.5KG
A1244-500GM
A1244-45KGBL
700009-910EA
1853.9
USD
700009-910
700009-912
700009-914
75810-506
Diammonium hydrogen orthophosphate ≥96.0%, granular NF
Diammonium hydrogen orthophosphate
Ammonium Phosphate, Granular, NF is used a buffering agent excipient.
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Formula:
(NH₄)₂HPO₄ MW: 132.06 g/mol Melting Pt: 185 Storage Temperature: Ambient |
MDL Number:
MFCD00010891 CAS Number: 7783-28-0 Merck Index: 13,00546 |
Specification Test Results
Assay [(NH₄)₂HPO₄] | 96.0-102.0 % |
pH of a 1 in 100 Solution | 7.6 - 8.2 |
Chloride (Cl) | max. 0.03 % |
Sulfates (SO₄) | max. 0.15 % |
Arsenic (As) | max. 3 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...