Ammoniated mercury ≥98.0%, Powder USP

Supplier: Spectrum Chemical Mfg. Corp.

AM168-500GM AM168-125GM AM168-2.5KG
95031-256EA 1482.2 USD
95031-256 95031-252 95031-324
Ammoniated mercury ≥98.0%, Powder USP
Ammoniated mercury

Ammoniated Mercury, Powder, USP is used to treat a variety of minor skin infections and other skin disorders. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Skull and crossbones;Health hazard UN1630

Formula: Hg(NH₂)Cl
Storage Temperature: Ambient
MDL Number: MFCD00016140
CAS Number: 10124-48-8
UN: 1630
ADR: 6.1,II

Order Now

Specification Test Results

Assay 98.0 - 100.5%
Residue After Ignition max. 0.2%
Mercurous Compounds max. 0.2%
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR