Acid red 27

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Amaranth, Sodium 3-hydroxy-4-((4-sulphonatonaphthalen-1-yl)diazenyl)naphthalene-2,7-disulphonate, Bordeaux S, Food Red No. 2, Azo Rubine S, Wool Red 40 F, Naphthol Red

AM105-100GM AM105-25GM
700009-990EA 136.4 USD
700009-990 700009-992
Acid red 27
Acid red 27

Amaranth, a genus of annual perennial plants, can be used as an additive to wheat flour. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.


GHS hazard : Exclamation mark

Formula: C₂₀H₁₄N₂O₁₀S₃
MW: 604.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004076
CAS Number: 915-67-3

Order Now

Specification Test Results

Absorption Max. (In Water) Approx. 521 NM

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR