Aluminum monostearate NF

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Dihydroxy(stearoyloxy)aluminum, Aluminum monostearate technical(Hard and soft aluminium mixture), Aluminum dihydroxide stearate

AL228-12KGBL AL228-500GM AL228-100GM
700004-966EA 13034.65 USD
700004-966 95031-740 95031-738
Aluminum monostearate NF
Aluminum monostearate

Aluminum Monostearate, NF is also known as Dihydroxyaluminum stearate. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: C₁₈H₃₇AlO₄
Storage Temperature: Ambient
MDL Number: MFCD00019932
CAS Number: 7047-84-9

Order Now

Specification Test Results

Assay (Al₂O₃) 14.5-16.5 %
Loss On Drying max.2.0 %
Arsenic (As) max. 4 ppm
Heavy Metals max. 50 Ug/G
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR