ɑ-Caryophyllene ≥93%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: ɑ-Humulene, Humulene

C2990-1ML C2990-5ML
10860-324EA 214.4 USD
10860-324 10860-326
ɑ-Caryophyllene ≥93%
alpha-Caryophyllene

GHS hazard : Exclamation mark

Formula: C₁₅H₂₄
MW: 204.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00042689
CAS Number: 6753-98-6

Order Now

Specification Test Results

Purity (GC) 93 %
Refractive Index (N20/D) 1.5020 - 1.5040

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR