Aloe vera (200:1), powder
Supplier: SPECTRUM CHEMICAL MFG CORP
Aloe Vera, 200:1, Powder - Spectrum's freeze dried aloe vera powder is easy to use. Just add 199 parts distilled or deionized water to 1 part aloe vera powder to reconstitute the oil derived from the Aloe Vera plant. It is known for its moisturizing and healing qualities and can be used in a wide range of personal care products. It is also a humectant, hair conditioners, and to relieve dandruff and dry scalp.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Storage Temperature:
Ambient |
MDL Number:
MFCD04118389 CAS Number: 85507-69-3 |
Specification Test Results
Moisture | max. 10 % |
Microbial Limits | max. 100 Cfu/G |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...