Almond oil NF, sweet
Supplier: Spectrum Chemical Mfg. Corp.
Almond Oil, Sweet, NF (AL215) is the fixed oil obtained by expression with hydraulic or expeller presses from the kernels of sweet almonds. It is a natural, versatile and effective emollient known for its stability and high level of the skin nourishing Linoleic Acid. It is a natural moisturizer and is suitable for many personal care products, cosmetics, conditioners, and fragrances. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Storage temperature - LIGHT SENSITIVE: Keep tightly closed in light-resistant containers.
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Storage Temperature:
Ambient |
MDL Number:
MFCD00130467 CAS Number: 8007-69-0 |
Specification Test Results
Specific Gravity @ 25°C | 0.910 - 0.915 |
Fatty Acid Composition | To Pass Test |
Sterol Composition | To Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...