5-(2,5-Dioxotetrahydrofuryl)-3-methyl-3-cyclohexene-1,2-dicarboxylic anhydride ≥95.0%
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
6-(2,5-Dioxotetrahydrofuran-3-yl)-4-methyl-7,7a-dihydroisobenzofuran-1,3(3aH,6H)-dione
D1574-25GM
D1574-100GM
10860-314EA
105.85
USD
10860-314
10860-316
5-(2,5-Dioxotetrahydrofuryl)-3-methyl-3-cyclohexene-1,2-dicarboxylic anhydride ≥95.0%
5-(2,5-Dioxotetrahydrofuryl)-3-methyl-3-cyclohexene-1,2-dicarboxylic anhydride
Storage temperature - MOISTURE SENSITIVE: Keep tightly closed. Store under inert gas.
GHS hazard : Exclamation mark
Precaución: Storage temperature - MOISTURE SENSITIVE: Keep tightly closed. Store under inert gas.
Formula:
C₁₃H₁₂O₆ MW: 264.23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00039141 CAS Number: 73003-90-4 |
Specification Test Results
Purity (Methanolysis Method) | 95.0 % min. |
Melting Point | 172.0 - 176.0 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...